Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01523954_m1
          See other LIPG GE Assays ›
          Gene Symbol:
          Lipg
          Gene Name:
          lipase, endothelial
          Gene Aliases:
          lipase
          Chromosome Location:
          Chr.18: 70903528 - 70924434 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01523954_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012741.1 NP_001012759.1 7 - 8 1166 108
          GenBank mRNA AY916123.1 - 7 - 8 1166 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001704.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lipg

          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to nutrient
          cell proliferation
          positive regulation of high-density lipoprotein particle clearance
          lipid catabolic process
          positive regulation of cholesterol transport
          high-density lipoprotein particle remodeling
          cholesterol homeostasis
          reverse cholesterol transport
          regulation of lipoprotein metabolic process
          phospholipid homeostasis
          molecular_function
          lipoprotein lipase activity
          phospholipase activity
          heparin binding
          phosphatidylcholine 1-acylhydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P151151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151151 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151151 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P151151 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337441 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4892 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37525 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805237 - -
          Affymetrix Rat Gene 1.0 ST Array 10805234 - -
          Affymetrix Rat Gene 1.0 ST Array 10805235 - -
          Affymetrix Rat Gene 1.0 ST Array 10805232 - -
          Affymetrix Rat Gene 1.0 ST Array 10805239 - -
          Affymetrix Rat Gene 1.0 ST Array 10805233 - -
          Affymetrix Rat Gene 1.0 ST Array 10805236 - -
          Affymetrix Rat Gene 1.0 ST Array 10805231 - -
          Affymetrix Rat Gene 1.0 ST Array 10805238 - -
          Affymetrix Rat Gene 1.1 ST Array 10805229 - -
          Rat Genome 230 2.0 Array 1390790_PM_a_at - -
          Rat Genome 230 2.0 Array 1390790_a_at - -
          Affymetrix Rat Clariom S Assay PSR1800011915.rn.1 TC1800001704.rn.1 RTA TC
          Rat Expression Set 230 1390790_a_at - -
          Rat Expression Set 230 1390790_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805229 - -
          Rat Genome U34 Set rc_AA964219_at - -
          Rat Genome U34 Set rc_AI236616_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651147 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline