Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01523993_m1
          See other MYO5B GE Assays ›
          Gene Symbol:
          Myo5b
          Gene Name:
          myosin Vb
          Gene Aliases:
          Myr6
          Chromosome Location:
          Chr.18: 70427103 - 70729985 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01523993_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myo5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017083.1 NP_058779.1 36 - 37 5005 87
          XM_006254908.2 XP_006254970.1 32 - 33 4458 87
          XM_008772085.1 XP_008770307.1 36 - 37 4968 87
          XM_008772086.1 XP_008770308.1 35 - 36 4887 87
          GenBank mRNA U60416.1 - 36 - 37 5005 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000815.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myo5b

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          protein transport
          vesicle-mediated transport
          endosome localization
          regulation of protein localization
          motor activity
          actin binding
          protein binding
          calmodulin binding
          ATP binding
          Rab GTPase binding
          ionotropic glutamate receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P339558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15811 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151146 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P339558 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P339558 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P339558 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P339558 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702154310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80138 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65891 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57894 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80015 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56667 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24020 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62732 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41898 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95819 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1592 - -
          Agilent Rat cDNA Microarray (G4105A) 702154310 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802672 - -
          Affymetrix Rat Gene 1.0 ST Array 10802662 - -
          Affymetrix Rat Gene 1.0 ST Array 10802656 - -
          Affymetrix Rat Gene 1.0 ST Array 10802655 - -
          Affymetrix Rat Gene 1.0 ST Array 10802663 - -
          Affymetrix Rat Gene 1.0 ST Array 10802670 - -
          Affymetrix Rat Gene 1.0 ST Array 10802666 - -
          Affymetrix Rat Gene 1.0 ST Array 10802654 - -
          Affymetrix Rat Gene 1.0 ST Array 10802668 - -
          Affymetrix Rat Gene 1.0 ST Array 10802683 - -
          Affymetrix Rat Gene 1.0 ST Array 10802671 - -
          Affymetrix Rat Gene 1.0 ST Array 10802675 - -
          Affymetrix Rat Gene 1.0 ST Array 10802678 - -
          Affymetrix Rat Gene 1.0 ST Array 10802682 - -
          Affymetrix Rat Gene 1.0 ST Array 10802657 - -
          Affymetrix Rat Gene 1.0 ST Array 10802680 - -
          Affymetrix Rat Gene 1.0 ST Array 10802674 - -
          Affymetrix Rat Gene 1.0 ST Array 10802658 - -
          Affymetrix Rat Gene 1.0 ST Array 10802686 - -
          Affymetrix Rat Gene 1.0 ST Array 10802669 - -
          Affymetrix Rat Gene 1.0 ST Array 10802684 - -
          Affymetrix Rat Gene 1.0 ST Array 10802659 - -
          Affymetrix Rat Gene 1.0 ST Array 10802690 - -
          Affymetrix Rat Gene 1.0 ST Array 10802676 - -
          Affymetrix Rat Gene 1.0 ST Array 10802661 - -
          Affymetrix Rat Gene 1.0 ST Array 10802664 - -
          Affymetrix Rat Gene 1.0 ST Array 10802651 - -
          Affymetrix Rat Gene 1.0 ST Array 10802677 - -
          Affymetrix Rat Gene 1.0 ST Array 10802667 - -
          Affymetrix Rat Gene 1.0 ST Array 10802660 - -
          Affymetrix Rat Gene 1.0 ST Array 10802689 - -
          Affymetrix Rat Gene 1.0 ST Array 10802681 - -
          Affymetrix Rat Gene 1.0 ST Array 10802688 - -
          Affymetrix Rat Gene 1.0 ST Array 10802673 - -
          Affymetrix Rat Gene 1.0 ST Array 10802685 - -
          Affymetrix Rat Gene 1.0 ST Array 10802687 - -
          Affymetrix Rat Gene 1.0 ST Array 10802652 - -
          Affymetrix Rat Gene 1.0 ST Array 10802653 - -
          Affymetrix Rat Gene 1.0 ST Array 10802679 - -
          Affymetrix Rat Gene 1.1 ST Array 10802650 - -
          Rat Genome 230 2.0 Array 1368355_at - -
          Rat Genome 230 2.0 Array 1368355_PM_at - -
          Affymetrix HT Rat Focus Array 1368355_at - -
          Affymetrix HT Rat Focus Array 1368355_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800005875.rn.1 TC1800000815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005879.rn.1 TC1800000815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005873.rn.1 TC1800000815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005878.rn.1 TC1800000815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005876.rn.1 TC1800000815.rn.1 RTA TC
          Rat Expression Set 230 1368355_at - -
          Rat Expression Set 230 1368355_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802650 - -
          Rat Genome U34 Set U60416_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21040479 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373346 - -
          RatRef-12 Expression BeadChip ILMN_51493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline