Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01524859_m1
          See other FLI1 GE Assays ›
          Gene Symbol:
          Fli1
          Gene Name:
          Fli-1 proto-oncogene, ETS transcription factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 33541932 - 33661094 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01524859_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fli1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017381.1 NP_001017381.1 8 - 9 1100 131
          XM_008766055.1 XP_008764277.1 7 - 8 1069 131
          GenBank mRNA AY972521.1 - 8 - 9 1100 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002192.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fli1

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          blood circulation
          organ morphogenesis
          cell differentiation
          megakaryocyte development
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19280 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17926 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17926 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17926 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17926 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74575 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28016 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72835 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37793 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10916002 - -
          Affymetrix Rat Gene 1.0 ST Array 10916005 - -
          Affymetrix Rat Gene 1.0 ST Array 10916011 - -
          Affymetrix Rat Gene 1.0 ST Array 10916010 - -
          Affymetrix Rat Gene 1.0 ST Array 10916003 - -
          Affymetrix Rat Gene 1.0 ST Array 10916001 - -
          Affymetrix Rat Gene 1.0 ST Array 10916004 - -
          Affymetrix Rat Gene 1.0 ST Array 10916006 - -
          Affymetrix Rat Gene 1.0 ST Array 10916007 - -
          Affymetrix Rat Gene 1.0 ST Array 10916008 - -
          Affymetrix Rat Gene 1.1 ST Array 10916000 - -
          Rat Genome 230 2.0 Array 1383199_PM_at - -
          Rat Genome 230 2.0 Array 1390914_at - -
          Rat Genome 230 2.0 Array 1377494_PM_at - -
          Rat Genome 230 2.0 Array 1383199_at - -
          Rat Genome 230 2.0 Array 1390914_PM_at - -
          Rat Genome 230 2.0 Array 1393249_PM_at - -
          Rat Genome 230 2.0 Array 1393249_at - -
          Rat Genome 230 2.0 Array 1377494_at - -
          Affymetrix HT Rat Focus Array 1377494_at - -
          Affymetrix HT Rat Focus Array 1390914_at - -
          Affymetrix HT Rat Focus Array 1377494_PM_at - -
          Affymetrix HT Rat Focus Array 1390914_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800017667.rn.1 TC0800002192.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017668.rn.1 TC0800002192.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017666.rn.1 TC0800002192.rn.1 RTA TC
          Rat Expression Set 230 1377494_PM_at - -
          Rat Expression Set 230 1377494_at - -
          Rat Expression Set 230 1390914_PM_at - -
          Rat Expression Set 230 1390914_at - -
          Affymetrix Rat Gene 1.0 ST Array 10916000 - -
          Rat Genome U34 Set rc_AI071474_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21663546 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651013 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline