Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01524948_m1
          See other NCAPD3 GE Assays ›
          Gene Symbol:
          Ncapd3
          Gene Name:
          non-SMC condensin II complex, subunit D3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 28075632 - 28145024 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01524948_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncapd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034000.1 NP_001029172.1 32 - 33 4348 64
          XM_006242735.2 XP_006242797.1 32 - 33 4650 64
          XM_006242736.2 XP_006242798.1 32 - 33 4301 64
          XM_008766053.1 XP_008764275.1 31 - 32 4725 64
          GenBank mRNA BC101928.1 - 32 - 33 4348 7939116 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000291.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ncapd3

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic chromosome condensation
          methylated histone binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29954 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11380 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11380 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11380 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21931 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11380 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89516 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908703 - -
          Affymetrix Rat Gene 1.0 ST Array 10908709 - -
          Affymetrix Rat Gene 1.0 ST Array 10908712 - -
          Affymetrix Rat Gene 1.0 ST Array 10908726 - -
          Affymetrix Rat Gene 1.0 ST Array 10908706 - -
          Affymetrix Rat Gene 1.0 ST Array 10908714 - -
          Affymetrix Rat Gene 1.0 ST Array 10908708 - -
          Affymetrix Rat Gene 1.0 ST Array 10908715 - -
          Affymetrix Rat Gene 1.0 ST Array 10908721 - -
          Affymetrix Rat Gene 1.0 ST Array 10908717 - -
          Affymetrix Rat Gene 1.0 ST Array 10908707 - -
          Affymetrix Rat Gene 1.0 ST Array 10908719 - -
          Affymetrix Rat Gene 1.0 ST Array 10908724 - -
          Affymetrix Rat Gene 1.0 ST Array 10908711 - -
          Affymetrix Rat Gene 1.0 ST Array 10908727 - -
          Affymetrix Rat Gene 1.0 ST Array 10908710 - -
          Affymetrix Rat Gene 1.0 ST Array 10908728 - -
          Affymetrix Rat Gene 1.0 ST Array 10908725 - -
          Affymetrix Rat Gene 1.0 ST Array 10908723 - -
          Affymetrix Rat Gene 1.0 ST Array 10908718 - -
          Affymetrix Rat Gene 1.0 ST Array 10908704 - -
          Affymetrix Rat Gene 1.0 ST Array 10908720 - -
          Affymetrix Rat Gene 1.0 ST Array 10908713 - -
          Affymetrix Rat Gene 1.0 ST Array 10908705 - -
          Affymetrix Rat Gene 1.0 ST Array 10908722 - -
          Affymetrix Rat Gene 1.0 ST Array 10908716 - -
          Affymetrix Rat Gene 1.1 ST Array 10908702 - -
          Rat Genome 230 2.0 Array 1391604_PM_at - -
          Rat Genome 230 2.0 Array 1391604_at - -
          Affymetrix HT Rat Focus Array 1391604_PM_at - -
          Affymetrix HT Rat Focus Array 1391604_at - -
          Affymetrix Rat Clariom S Assay PSR0800001950.rn.1 TC0800000291.rn.1 RTA TC
          Rat Expression Set 230 1391604_PM_at - -
          Rat Expression Set 230 1391604_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908702 - -
          Rat Genome U34 Set rc_AA943049_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349604 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367666 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-kvkbn:80/100.66.78.154:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline