Search
Search
| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007661.2 | NP_001007662.1 | 3 - 4 | 370 | 60 | |
| GenBank mRNA | BC064031.1 | - | 3 - 4 | 374 | 6887845 | 60 |
| FQ213324.1 | - | 2 - 3 | 519 | 60 | ||
Set Membership: |
|