Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01525092_m1
          See other FOXP3 GE Assays ›
          Gene Symbol:
          Foxp3
          Gene Name:
          forkhead box P3
          Gene Aliases:
          RGD1562112
          Chromosome Location:
          Chr.X: 15753175 - 15768643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01525092_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108250.1 NP_001101720.1 7 - 8 882 72
          XM_006256731.2 XP_006256793.1 6 - 7 945 72
          GenBank mRNA AB232988.1 - 7 - 8 906 72
          AB232989.1 - 6 - 7 824 72
          AB232990.1 - 7 - 8 906 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001510.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          B cell homeostasis
          cytokine production
          myeloid cell homeostasis
          CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
          T cell mediated immunity
          tolerance induction
          tolerance induction to self antigen
          regulation of immunoglobulin production
          positive regulation of T cell tolerance induction
          positive regulation of T cell anergy
          negative regulation of chronic inflammatory response
          negative regulation of T cell cytokine production
          positive regulation of peripheral T cell tolerance induction
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          biological_process
          negative regulation of cell proliferation
          response to virus
          positive regulation of gene expression
          negative regulation of histone deacetylation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interferon-gamma production
          negative regulation of interleukin-10 production
          negative regulation of interleukin-17 production
          negative regulation of interleukin-2 production
          negative regulation of interleukin-4 production
          negative regulation of interleukin-5 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-4 production
          negative regulation of CREB transcription factor activity
          positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
          positive regulation of transforming growth factor beta1 production
          positive regulation of immature T cell proliferation in thymus
          positive regulation of histone acetylation
          negative regulation of histone acetylation
          negative regulation of cytokine biosynthetic process
          T cell activation
          negative regulation of T cell proliferation
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulatory T cell differentiation
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of interleukin-2 biosynthetic process
          positive regulation of regulatory T cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of isotype switching to IgE isotypes
          regulation of isotype switching to IgG isotypes
          negative regulation of lymphocyte proliferation
          negative regulation of cytokine secretion
          negative regulation of inflammatory response
          negative regulation of immune response
          T cell receptor signaling pathway
          negative regulation of T-helper 17 cell differentiation
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          molecular_function
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription corepressor activity
          histone acetyltransferase binding
          protein homodimerization activity
          histone deacetylase binding
          sequence-specific DNA binding
          metal ion binding
          NFAT protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0X00009752.rn.1 TC0X00001510.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline