Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212458.1 | NP_997623.1 | 6 - 7 | 935 | 114 | |
XM_006255972.1 | XP_006256034.1 | 6 - 7 | 788 | 114 | ||
XM_006255973.1 | XP_006256035.1 | 5 - 6 | 689 | 114 | ||
XM_006255974.1 | XP_006256036.1 | 6 - 7 | 877 | 114 | ||
XM_006255975.1 | XP_006256037.1 | 6 - 7 | 1178 | 114 | ||
GenBank mRNA | BC085852.1 | - | 6 - 7 | 881 | 7189891 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |