Search
Search
| Gene Symbol: Ezr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019357.1 | NP_062230.1 | 7 - 8 | 1274 | 87 | |
| GenBank mRNA | AF450298.1 | - | 7 - 8 | 873 | 87 | |
| AY428869.1 | - | 7 - 8 | 859 | 87 | ||
| BC081958.1 | - | 7 - 8 | 1274 | 7124292 | 87 | |
Set Membership: |
|