Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01526363_m1
          See other WTAP GE Assays ›
          Gene Symbol:
          Wtap
          Gene Name:
          Wilms tumor 1 associated protein
          Gene Aliases:
          RGD1563824
          Chromosome Location:
          Chr.1: 47942500 - 47967633 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01526363_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wtap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113542.1 NP_001107014.1 5 - 6 466 64
          NM_001113543.1 NP_001107015.1 5 - 6 466 64
          NM_001113544.1 NP_001107016.1 5 - 6 400 64
          XM_006227892.2 XP_006227954.1 5 - 6 446 64
          XM_006227893.2 XP_006227955.1 5 - 6 624 64
          GenBank mRNA CB613741.1 - 5 - 6 395 64
          CB613959.1 - 5 - 6 400 64
          CK478322.1 - 5 - 6 466 64
          CV108433.1 - 5 - 6 463 64
          CX570823.1 - 1 - 2 112 64
          FQ228682.1 - 5 - 6 505 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000501.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA methylation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15181 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P595828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112728 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P595828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15181 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P595828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15181 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P595828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15181 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15181 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P285915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35365 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702942 - -
          Affymetrix Rat Gene 1.0 ST Array 10702943 - -
          Affymetrix Rat Gene 1.0 ST Array 10702941 - -
          Affymetrix Rat Gene 1.0 ST Array 10702944 - -
          Affymetrix Rat Gene 1.0 ST Array 10702937 - -
          Affymetrix Rat Gene 1.0 ST Array 10702940 - -
          Affymetrix Rat Gene 1.0 ST Array 10702939 - -
          Affymetrix Rat Gene 1.0 ST Array 10702938 - -
          Affymetrix Rat Gene 1.1 ST Array 10702936 - -
          Rat Genome 230 2.0 Array 1379249_PM_at - -
          Rat Genome 230 2.0 Array 1382175_at - -
          Rat Genome 230 2.0 Array 1379249_at - -
          Rat Genome 230 2.0 Array 1382175_PM_at - -
          Affymetrix HT Rat Focus Array 1382175_PM_at - -
          Affymetrix HT Rat Focus Array 1382175_at - -
          Affymetrix Rat Clariom S Assay PSR0100003128.rn.1 TC0100000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003119.rn.1 TC0100000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003127.rn.1 TC0100000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003120.rn.1 TC0100000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003129.rn.1 TC0100000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003122.rn.1 TC0100000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003130.rn.1 TC0100000501.rn.1 RTA TC
          Rat Expression Set 230 1379249_PM_at - -
          Rat Expression Set 230 1382175_PM_at - -
          Rat Expression Set 230 1382175_at - -
          Rat Expression Set 230 1379249_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702936 - -
          Rat Genome U34 Set rc_AA900400_at - -
          Rat Genome U34 Set rc_AI176435_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22333267 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline