Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100575.1 | NP_001094045.1 | 3 - 4 | 348 | 60 | |
XM_006254138.2 | XP_006254200.1 | 4 - 5 | 2131 | 60 | ||
GenBank mRNA | BC085813.1 | - | 3 - 4 | 431 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |