Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01526680_m1
          See other DMTF1 GE Assays ›
          Gene Symbol:
          Dmtf1
          Gene Name:
          cyclin D binding myb-like transcription factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 21862313 - 21902798 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01526680_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dmtf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053693.1 NP_446145.1 7 - 8 796 143
          XM_006235993.1 XP_006236055.1 6 - 7 648 143
          GenBank mRNA BC081880.1 - 7 - 8 796 7108159 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000231.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dmtf1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116079 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119667 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P213482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035109 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P213481 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119667 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035109 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P213481 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119667 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035109 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P213481 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119667 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P278639 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410232 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P111921 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191903 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853277 - -
          Affymetrix Rat Gene 1.0 ST Array 10853275 - -
          Affymetrix Rat Gene 1.0 ST Array 10853281 - -
          Affymetrix Rat Gene 1.0 ST Array 10853282 - -
          Affymetrix Rat Gene 1.0 ST Array 10853289 - -
          Affymetrix Rat Gene 1.0 ST Array 10853274 - -
          Affymetrix Rat Gene 1.0 ST Array 10853279 - -
          Affymetrix Rat Gene 1.0 ST Array 10853278 - -
          Affymetrix Rat Gene 1.0 ST Array 10853286 - -
          Affymetrix Rat Gene 1.0 ST Array 10853280 - -
          Affymetrix Rat Gene 1.0 ST Array 10853276 - -
          Affymetrix Rat Gene 1.0 ST Array 10800338 - -
          Affymetrix Rat Gene 1.0 ST Array 10853285 - -
          Affymetrix Rat Gene 1.1 ST Array 10853273 - -
          Affymetrix Rat Gene 1.1 ST Array 10800337 - -
          Rat Genome 230 2.0 Array 1375666_at - -
          Rat Genome 230 2.0 Array 1375666_PM_at - -
          Affymetrix HT Rat Focus Array 1375666_at - -
          Affymetrix HT Rat Focus Array 1375666_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400001629.rn.1 TC0400000231.rn.1 RTA TC
          Rat Expression Set 230 1375666_PM_at - -
          Rat Expression Set 230 1375666_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853273 - -
          Rat Genome U34 Set rc_AA866459_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20923270 - -
          ABI Rat Genome Survey Microarray 21980151 - -
          ABI Rat Genome Survey Microarray 21991836 - -
          ABI Rat Genome Survey Microarray 22394311 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376429 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375855 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline