Search
Search

Rundc3b| Gene Symbol: Rundc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047116.1 | NP_001040581.1 | 4 - 5 | 700 | 149 | |
| XM_006236003.2 | XP_006236065.1 | 4 - 5 | 1744 | 149 | ||
| GenBank mRNA | BC107563.1 | - | 4 - 5 | 700 | 7938579 | 149 |
Set Membership: |
|