Search
Search
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023093.1 | NP_075581.1 | 2 - 3 | 188 | 68 | |
| GenBank mRNA | BC061830.1 | - | 2 - 3 | 188 | 5599635 | 68 |
| J05266.1 | - | 2 - 3 | 109 | 68 | ||
Set Membership: |
|