Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527043_m1
          See other PIAS2 GE Assays ›
          Gene Symbol:
          Pias2
          Gene Name:
          protein inhibitor of activated STAT, 2
          Gene Aliases:
          Miz1
          Chromosome Location:
          Chr.18: 73479863 - 73524956 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527043_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pias2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053337.1 NP_445789.1 693 82
          GenBank mRNA AF044058.1 - 693 82
          BC078775.1 - 692 7111628 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000853.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pias2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          spermatogenesis
          metabolic process
          response to organic substance
          protein sumoylation
          androgen receptor signaling pathway
          response to testosterone
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of dendrite morphogenesis
          regulation of androgen receptor signaling pathway
          negative regulation of androgen receptor signaling pathway
          DNA binding
          protein binding
          transcription factor binding
          zinc ion binding
          ligase activity
          SUMO transferase activity
          protein domain specific binding
          estrogen receptor binding
          ubiquitin protein ligase binding
          glucocorticoid receptor binding
          androgen receptor binding
          SUMO ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P377245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158143 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P377245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P377245 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P377245 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P377245 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701745418 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68505 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93911 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68027 - -
          Agilent Rat cDNA Microarray (G4105A) 701745418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802792 - -
          Affymetrix Rat Gene 1.0 ST Array 10802783 - -
          Affymetrix Rat Gene 1.0 ST Array 10802788 - -
          Affymetrix Rat Gene 1.0 ST Array 10802786 - -
          Affymetrix Rat Gene 1.0 ST Array 10802793 - -
          Affymetrix Rat Gene 1.0 ST Array 10802787 - -
          Affymetrix Rat Gene 1.0 ST Array 10802784 - -
          Affymetrix Rat Gene 1.0 ST Array 10802790 - -
          Affymetrix Rat Gene 1.0 ST Array 10802789 - -
          Affymetrix Rat Gene 1.0 ST Array 10802791 - -
          Affymetrix Rat Gene 1.0 ST Array 10802785 - -
          Affymetrix Rat Gene 1.1 ST Array 10802782 - -
          Rat Genome 230 2.0 Array 1387299_PM_at - -
          Rat Genome 230 2.0 Array 1394413_at - -
          Rat Genome 230 2.0 Array 1394413_PM_at - -
          Rat Genome 230 2.0 Array 1387299_at - -
          Affymetrix HT Rat Focus Array 1387299_PM_at - -
          Affymetrix HT Rat Focus Array 1387299_at - -
          Affymetrix Rat Clariom S Assay PSR1800006150.rn.1 TC1800000853.rn.1 RTA TC
          Rat Expression Set 230 1387299_PM_at - -
          Rat Expression Set 230 1387299_at - -
          Rat Expression Set 230 1394413_at - -
          Rat Expression Set 230 1394413_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802782 - -
          Rat Genome U34 Set AF044058_at - -
          Rat Genome U34 Set rc_AI145852_at - -
          Rat Genome U34 Set rc_AI137350_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21367020 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370272 - -
          RatRef-12 Expression BeadChip ILMN_63967 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline