Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527137_m1
          See other TIMELESS GE Assays ›
          Gene Symbol:
          Timeless
          Gene Name:
          timeless circadian clock
          Gene Aliases:
          Tim
          Chromosome Location:
          Chr.7: 2643288 - 2667329 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527137_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timeless
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031340.1 NP_112630.1 26 - 27 3331 78
          GenBank mRNA AB019576.1 - 26 - 27 3331 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000024.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timeless

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          branching involved in ureteric bud morphogenesis
          kidney development
          morphogenesis of an epithelium
          transcription, DNA-templated
          cellular response to DNA damage stimulus
          mitotic nuclear division
          circadian rhythm
          lung development
          regulation of cell proliferation
          regulation of circadian rhythm
          positive regulation of circadian rhythm
          cell cycle phase transition
          negative regulation of transcription, DNA-templated
          branching morphogenesis of an epithelial tube
          cell division
          protein binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P810736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P810736 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P810736 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P810736 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P810736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81762 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76921 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10893054 - -
          Affymetrix Rat Gene 1.0 ST Array 10893061 - -
          Affymetrix Rat Gene 1.0 ST Array 10893040 - -
          Affymetrix Rat Gene 1.0 ST Array 10893055 - -
          Affymetrix Rat Gene 1.0 ST Array 10893062 - -
          Affymetrix Rat Gene 1.0 ST Array 10893044 - -
          Affymetrix Rat Gene 1.0 ST Array 10893047 - -
          Affymetrix Rat Gene 1.0 ST Array 10893041 - -
          Affymetrix Rat Gene 1.0 ST Array 10893058 - -
          Affymetrix Rat Gene 1.0 ST Array 10893042 - -
          Affymetrix Rat Gene 1.0 ST Array 10893051 - -
          Affymetrix Rat Gene 1.0 ST Array 10893057 - -
          Affymetrix Rat Gene 1.0 ST Array 10893045 - -
          Affymetrix Rat Gene 1.0 ST Array 10893038 - -
          Affymetrix Rat Gene 1.0 ST Array 10893053 - -
          Affymetrix Rat Gene 1.0 ST Array 10893056 - -
          Affymetrix Rat Gene 1.0 ST Array 10893052 - -
          Affymetrix Rat Gene 1.0 ST Array 10893048 - -
          Affymetrix Rat Gene 1.0 ST Array 10893043 - -
          Affymetrix Rat Gene 1.0 ST Array 10893037 - -
          Affymetrix Rat Gene 1.0 ST Array 10893049 - -
          Affymetrix Rat Gene 1.0 ST Array 10893046 - -
          Affymetrix Rat Gene 1.0 ST Array 10893039 - -
          Affymetrix Rat Gene 1.0 ST Array 10893050 - -
          Affymetrix Rat Gene 1.0 ST Array 10893063 - -
          Affymetrix Rat Gene 1.0 ST Array 10893036 - -
          Affymetrix Rat Gene 1.0 ST Array 10893060 - -
          Affymetrix Rat Gene 1.0 ST Array 10893059 - -
          Affymetrix Rat Gene 1.1 ST Array 10893035 - -
          Rat Genome 230 2.0 Array 1368522_PM_at - -
          Rat Genome 230 2.0 Array 1368522_at - -
          Affymetrix HT Rat Focus Array 1368522_PM_at - -
          Affymetrix HT Rat Focus Array 1368522_at - -
          Affymetrix Rat Clariom S Assay PSR0700000298.rn.1 TC0700000024.rn.1 RTA TC
          Rat Expression Set 230 1368522_at - -
          Rat Expression Set 230 1368522_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10893035 - -
          Rat Genome U34 Set AB019576_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21998244 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370013 - -
          RatRef-12 Expression BeadChip ILMN_54777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline