Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527268_m1
          See other RGD1563060 GE Assays ›
          Gene Symbol:
          RGD1563060
          Gene Name:
          similar to AVLV472
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 38307889 - 38329780 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527268_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1563060
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106149.1 NP_001099619.1 3 - 4 378 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001367.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147743 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160338 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160338 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P344791 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20102 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10804296 - -
          Affymetrix Rat Gene 1.0 ST Array 10804295 - -
          Affymetrix Rat Gene 1.0 ST Array 10804294 - -
          Affymetrix Rat Gene 1.0 ST Array 10804293 - -
          Affymetrix Rat Gene 1.1 ST Array 10804292 - -
          Rat Genome 230 2.0 Array 1378398_at - -
          Rat Genome 230 2.0 Array 1378398_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800009859.rn.1 TC1800001367.rn.1 RTA TC
          Rat Expression Set 230 1378398_PM_at - -
          Rat Expression Set 230 1378398_at - -
          Affymetrix Rat Gene 1.0 ST Array 10804292 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21903002 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374758 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline