Search
Search
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766803.1 | XP_008765025.1 | 19 - 20 | 2616 | 61 | |
| XM_008766804.1 | XP_008765026.1 | 18 - 19 | 3426 | 61 | ||
| XM_217309.8 | XP_217309.6 | 19 - 20 | 3245 | 61 | ||
Set Membership: |
|