Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527840_m1
          See other HPRT1 GE Assays ›
          Gene Symbol:
          Hprt1
          Gene Name:
          hypoxanthine phosphoribosyltransferase 1
          Gene Aliases:
          Hgprtase, Hprt
          Chromosome Location:
          Chr.X: 158196640 - 158228815 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527840_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (2)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hprt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012583.2 NP_036715.1 8 - 9 673 64
          GenBank mRNA BC098629.1 - 8 - 9 673 7386548 64
          FQ212770.1 - 257 64
          FQ215291.1 - 257 64
          FQ215779.1 - 257 64
          FQ216318.1 - 257 64
          FQ219798.1 - 257 64
          FQ219974.1 - 257 64
          FQ220004.1 - 257 64
          FQ229708.1 - 257 64
          FQ230307.1 - 257 64
          FQ234297.1 - 257 64
          M63983.1 - 8 - 9 705 64
          M86443.1 - 433 64
          S79292.1 - 8 - 9 705 64
          X62085.1 - 633 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Endogenous Controls Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Inflammation Rat Phosphodiesterase Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Rat Phosphodiesterase Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hprt1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          purine nucleotide biosynthetic process
          purine ribonucleoside salvage
          adenine salvage
          guanine salvage
          spermatogenesis
          brain development
          grooming behavior
          locomotory behavior
          cytolysis
          striatum development
          cerebral cortex neuron differentiation
          central nervous system neuron development
          GMP salvage
          IMP salvage
          cellular response to insulin stimulus
          dopamine metabolic process
          hypoxanthine salvage
          positive regulation of dopamine metabolic process
          GMP catabolic process
          IMP metabolic process
          adenine metabolic process
          hypoxanthine metabolic process
          lymphocyte proliferation
          dendrite morphogenesis
          protein homotetramerization
          nucleotide binding
          magnesium ion binding
          hypoxanthine phosphoribosyltransferase activity
          identical protein binding
          protein homodimerization activity
          guanine phosphoribosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156445 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11257 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P575340 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1058445 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365155 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700253328 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58638 - -
          Agilent Rat cDNA Microarray (G4105A) 700253328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791546 - -
          Affymetrix Rat Gene 1.0 ST Array 10935476 - -
          Affymetrix Rat Gene 1.0 ST Array 10935478 - -
          Affymetrix Rat Gene 1.0 ST Array 10935477 - -
          Affymetrix Rat Gene 1.0 ST Array 10935479 - -
          Affymetrix Rat Gene 1.0 ST Array 10935480 - -
          Affymetrix Rat Gene 1.1 ST Array 10791545 - -
          Affymetrix Rat Gene 1.1 ST Array 10935475 - -
          Rat Genome 230 2.0 Array 1370280_PM_at - -
          Rat Genome 230 2.0 Array 1370280_at - -
          Affymetrix HT Rat Focus Array 1370280_PM_at - -
          Affymetrix HT Rat Focus Array 1370280_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016713.rn.1 TC0X00002660.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016714.rn.1 TC0X00002660.rn.1 RTA TC
          Rat Expression Set 230 1370280_at - -
          Rat Expression Set 230 1370280_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935475 - -
          Rat Genome U34 Set M63983_s_at - -
          Rat Genome U34 Set M63983_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336170 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367708 - -
          RatRef-12 Expression BeadChip ILMN_67287 - -

          Back To Top
           

          Citations (2)

          Presence, actions, and regulation of myostatin in rat uterus and myometrial cells.
          Authors
          Ciarmela P, Wiater E, Smith SM, Vale W
          Journal
          Endocrinology - Page No 906 (Sun Feb 01 00:00:00 EST 2009)
          Volume (Year)
          150 (2009)
          Product Usage
          -
          Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
          Authors
          Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
          Journal
          J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 EDT 2009)
          Volume (Year)
          201 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline