Search
Search
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014107.1 | NP_001014129.1 | 4 - 5 | 396 | 71 | |
| XM_006257631.2 | XP_006257693.1 | 3 - 4 | 442 | 71 | ||
| XM_006257632.2 | XP_006257694.1 | 4 - 5 | 481 | 71 | ||
| XM_006257633.2 | XP_006257695.1 | 3 - 4 | 439 | 71 | ||
| XM_006257634.2 | XP_006257696.1 | 3 - 4 | 439 | 71 | ||
| GenBank mRNA | BC086521.1 | - | 4 - 5 | 396 | 7302680 | 71 |
Set Membership: |
|