Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527980_m1
          See other PTCH1 GE Assays ›
          Gene Symbol:
          Ptch1
          Gene Name:
          patched 1
          Gene Aliases:
          Ptch, Ptch2
          Chromosome Location:
          Chr.17: 1032242 - 1085885 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527980_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptch1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053566.1 NP_446018.1 22 - 23 3765 70
          GenBank mRNA AY357891.1 - 22 - 23 3765 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001174.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptch1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          cell fate determination
          neural tube formation
          neural tube closure
          heart morphogenesis
          signal transduction
          smoothened signaling pathway
          regulation of mitotic cell cycle
          pattern specification process
          brain development
          negative regulation of cell proliferation
          epidermis development
          regulation of smoothened signaling pathway
          response to mechanical stimulus
          organ morphogenesis
          dorsal/ventral pattern formation
          epidermal cell fate specification
          response to chlorate
          positive regulation of cholesterol efflux
          response to organic cyclic compound
          protein processing
          spinal cord motor neuron differentiation
          neural tube patterning
          dorsal/ventral neural tube patterning
          neural plate axis specification
          embryonic limb morphogenesis
          mammary gland development
          response to estradiol
          response to retinoic acid
          regulation of protein localization
          limb morphogenesis
          hindlimb morphogenesis
          regulation of growth
          negative regulation of multicellular organism growth
          regulation of cell proliferation
          response to drug
          glucose homeostasis
          negative regulation of sequence-specific DNA binding transcription factor activity
          keratinocyte proliferation
          positive regulation of epidermal cell differentiation
          negative regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          embryonic organ development
          negative regulation of epithelial cell proliferation
          negative regulation of cell division
          pharyngeal system development
          mammary gland duct morphogenesis
          mammary gland epithelial cell differentiation
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          cell differentiation involved in kidney development
          somite development
          cellular response to cholesterol
          renal system development
          cell proliferation involved in metanephros development
          protein targeting to plasma membrane
          molecular_function
          patched binding
          smoothened binding
          hedgehog receptor activity
          heparin binding
          cholesterol binding
          cyclin binding
          protein complex binding
          hedgehog family protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86127 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147263 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147263 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147263 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19684 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15776 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701750422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19182 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19174 - -
          Agilent Rat cDNA Microarray (G4105A) 701750422 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10793534 - -
          Affymetrix Rat Gene 1.0 ST Array 10793544 - -
          Affymetrix Rat Gene 1.0 ST Array 10793551 - -
          Affymetrix Rat Gene 1.0 ST Array 10793530 - -
          Affymetrix Rat Gene 1.0 ST Array 10793552 - -
          Affymetrix Rat Gene 1.0 ST Array 10793538 - -
          Affymetrix Rat Gene 1.0 ST Array 10793542 - -
          Affymetrix Rat Gene 1.0 ST Array 10793531 - -
          Affymetrix Rat Gene 1.0 ST Array 10793543 - -
          Affymetrix Rat Gene 1.0 ST Array 10793533 - -
          Affymetrix Rat Gene 1.0 ST Array 10793547 - -
          Affymetrix Rat Gene 1.0 ST Array 10793535 - -
          Affymetrix Rat Gene 1.0 ST Array 10793541 - -
          Affymetrix Rat Gene 1.0 ST Array 10793548 - -
          Affymetrix Rat Gene 1.0 ST Array 10793540 - -
          Affymetrix Rat Gene 1.0 ST Array 10793537 - -
          Affymetrix Rat Gene 1.0 ST Array 10793536 - -
          Affymetrix Rat Gene 1.0 ST Array 10793545 - -
          Affymetrix Rat Gene 1.0 ST Array 10793539 - -
          Affymetrix Rat Gene 1.0 ST Array 10793546 - -
          Affymetrix Rat Gene 1.0 ST Array 10793549 - -
          Affymetrix Rat Gene 1.0 ST Array 10793550 - -
          Affymetrix Rat Gene 1.0 ST Array 10793532 - -
          Affymetrix Rat Gene 1.1 ST Array 10793528 - -
          Rat Genome 230 2.0 Array 1398614_at - -
          Rat Genome 230 2.0 Array 1398614_PM_at - -
          Affymetrix HT Rat Focus Array 1398614_at - -
          Affymetrix HT Rat Focus Array 1398614_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700008063.rn.1 TC1700001174.rn.1 RTA TC
          Rat Expression Set 230 1398614_PM_at - -
          Rat Expression Set 230 1398614_at - -
          Affymetrix Rat Gene 1.0 ST Array 10793528 - -
          Rat Genome U34 Set AF079162_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21579117 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371138 - -
          RatRef-12 Expression BeadChip ILMN_60929 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline