Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528101_m1
          See other STAG3 GE Assays ›
          Gene Symbol:
          Stag3
          Gene Name:
          stromal antigen 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 19599736 - 19630775 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528101_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stag3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053730.1 NP_446182.1 32 - 33 3734 84
          XM_006249020.2 XP_006249082.1 32 - 33 3902 84
          GenBank mRNA AY027880.1 - 32 - 33 3734 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000337.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stag3

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s)

          chromosome segregation
          meiotic nuclear division
          synapsis
          protein localization to chromosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P412236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62443 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412236 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412236 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P412236 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73733 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18532 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1387 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757294 - -
          Affymetrix Rat Gene 1.0 ST Array 10757298 - -
          Affymetrix Rat Gene 1.0 ST Array 10757295 - -
          Affymetrix Rat Gene 1.0 ST Array 10757286 - -
          Affymetrix Rat Gene 1.0 ST Array 10757270 - -
          Affymetrix Rat Gene 1.0 ST Array 10757277 - -
          Affymetrix Rat Gene 1.0 ST Array 10757271 - -
          Affymetrix Rat Gene 1.0 ST Array 10757268 - -
          Affymetrix Rat Gene 1.0 ST Array 10757279 - -
          Affymetrix Rat Gene 1.0 ST Array 10757284 - -
          Affymetrix Rat Gene 1.0 ST Array 10757282 - -
          Affymetrix Rat Gene 1.0 ST Array 10757276 - -
          Affymetrix Rat Gene 1.0 ST Array 10757273 - -
          Affymetrix Rat Gene 1.0 ST Array 10757281 - -
          Affymetrix Rat Gene 1.0 ST Array 10757297 - -
          Affymetrix Rat Gene 1.0 ST Array 10757269 - -
          Affymetrix Rat Gene 1.0 ST Array 10757266 - -
          Affymetrix Rat Gene 1.0 ST Array 10757272 - -
          Affymetrix Rat Gene 1.0 ST Array 10757283 - -
          Affymetrix Rat Gene 1.0 ST Array 10757265 - -
          Affymetrix Rat Gene 1.0 ST Array 10757288 - -
          Affymetrix Rat Gene 1.0 ST Array 10757289 - -
          Affymetrix Rat Gene 1.0 ST Array 10757292 - -
          Affymetrix Rat Gene 1.0 ST Array 10757280 - -
          Affymetrix Rat Gene 1.0 ST Array 10757293 - -
          Affymetrix Rat Gene 1.0 ST Array 10757296 - -
          Affymetrix Rat Gene 1.0 ST Array 10757291 - -
          Affymetrix Rat Gene 1.0 ST Array 10757267 - -
          Affymetrix Rat Gene 1.0 ST Array 10757275 - -
          Affymetrix Rat Gene 1.0 ST Array 10757274 - -
          Affymetrix Rat Gene 1.0 ST Array 10757290 - -
          Affymetrix Rat Gene 1.0 ST Array 10757285 - -
          Affymetrix Rat Gene 1.0 ST Array 10757278 - -
          Affymetrix Rat Gene 1.0 ST Array 10757287 - -
          Affymetrix Rat Gene 1.1 ST Array 10757264 - -
          Rat Genome 230 2.0 Array 1368613_PM_at - -
          Rat Genome 230 2.0 Array 1368613_at - -
          Affymetrix HT Rat Focus Array 1368613_PM_at - -
          Affymetrix HT Rat Focus Array 1368613_at - -
          Affymetrix Rat Clariom S Assay PSR1200002781.rn.1 TC1200000337.rn.1 RTA TC
          Rat Expression Set 230 1368613_PM_at - -
          Rat Expression Set 230 1368613_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757264 - -
          Rat Genome U34 Set rc_AI058833_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21604935 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370743 - -
          RatRef-12 Expression BeadChip ILMN_65290 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline