Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528294_g1
          See other SUV39H1 GE Assays ›
          Gene Symbol:
          Suv39h1
          Gene Name:
          suppressor of variegation 3-9 homolog 1 (Drosophila)
          Gene Aliases:
          RGD1565028
          Chromosome Location:
          Chr.X: 15171282 - 15185191 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528294_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Suv39h1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106956.1 NP_001100426.1 5 - 6 1560 81
          XM_006256703.2 XP_006256765.1 5 - 6 1776 81
          XM_006256704.2 XP_006256766.1 5 - 6 1352 81
          XM_006256705.2 XP_006256767.1 5 - 6 1353 81
          GenBank mRNA BC160871.1 - 5 - 6 1127 7364965 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000128.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing at rDNA
          biological_process
          histone H3-K9 methylation
          molecular_function
          zinc ion binding
          histone methyltransferase activity (H3-K9 specific)
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21231 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116989 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116981 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116989 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116981 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21231 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452731 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21231 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10932456 - -
          Affymetrix Rat Gene 1.0 ST Array 10940014 - -
          Affymetrix Rat Gene 1.0 ST Array 10932458 - -
          Affymetrix Rat Gene 1.0 ST Array 10940015 - -
          Affymetrix Rat Gene 1.0 ST Array 10932457 - -
          Affymetrix Rat Gene 1.0 ST Array 10932455 - -
          Affymetrix Rat Gene 1.1 ST Array 10932453 - -
          Affymetrix Rat Gene 1.1 ST Array 10940013 - -
          Rat Genome 230 2.0 Array 1375981_PM_a_at - -
          Rat Genome 230 2.0 Array 1375981_a_at - -
          Affymetrix HT Rat Focus Array 1375981_PM_a_at - -
          Affymetrix HT Rat Focus Array 1375981_a_at - -
          Affymetrix Rat Clariom S Assay PSR0X00000953.rn.1 TC0X00000128.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000957.rn.1 TC0X00000128.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000954.rn.1 TC0X00000128.rn.1 RTA TC
          Rat Expression Set 230 1375981_a_at - -
          Rat Expression Set 230 1375981_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10932453 - -
          Affymetrix Rat Gene 1.0 ST Array 10940013 - -
          Rat Genome U34 Set rc_AI011808_at - -
          Rat Genome U34 Set rc_AI011808_g_at - -
          Rat Genome U34 Set rc_AI177850_at - -
          Rat Genome U34 Set rc_AA858468_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21877754 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline