Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528452_g1
          See other ZKSCAN3 GE Assays ›
          Gene Symbol:
          Zkscan3
          Gene Name:
          zinc finger with KRAB and SCAN domains 3
          Gene Aliases:
          Zfp307
          Chromosome Location:
          Chr.17: 45247733 - 45263422 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528452_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zkscan3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012053.2 NP_001012053.2 4 - 5 716 136
          XM_006254096.2 XP_006254158.1 5 - 6 1546 136
          XM_006254099.2 XP_006254161.1 4 - 5 1043 136
          XM_006254101.2 XP_006254163.1 5 - 6 840 136
          XM_008771720.1 XP_008769942.1 4 - 5 6118 136
          XM_008771721.1 XP_008769943.1 4 - 5 6301 136
          XM_008771723.1 XP_008769945.1 5 - 6 6044 136
          XM_008771724.1 XP_008769946.1 5 - 6 5926 136
          XM_008771725.1 XP_008769947.1 5 - 6 6109 136
          XM_008771726.1 XP_008769948.1 5 - 6 1023 136
          GenBank mRNA BC089847.1 - 4 - 5 719 7373658 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000694.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zkscan3

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          lysosome organization
          negative regulation of autophagy
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of cellular senescence
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          nucleic acid binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094781 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P163131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163131 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P163131 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094781 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P163131 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P636608 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76089 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795341 - -
          Affymetrix Rat Gene 1.0 ST Array 10795338 - -
          Affymetrix Rat Gene 1.0 ST Array 10795336 - -
          Affymetrix Rat Gene 1.0 ST Array 10795337 - -
          Affymetrix Rat Gene 1.0 ST Array 10795343 - -
          Affymetrix Rat Gene 1.0 ST Array 10795340 - -
          Affymetrix Rat Gene 1.0 ST Array 10795339 - -
          Affymetrix Rat Gene 1.0 ST Array 10795342 - -
          Affymetrix Rat Gene 1.1 ST Array 10795335 - -
          Rat Genome 230 2.0 Array 1398573_at - -
          Rat Genome 230 2.0 Array 1398573_PM_at - -
          Rat Genome 230 2.0 Array 1382744_PM_at - -
          Rat Genome 230 2.0 Array 1382744_at - -
          Affymetrix HT Rat Focus Array 1398573_at - -
          Affymetrix HT Rat Focus Array 1382744_PM_at - -
          Affymetrix HT Rat Focus Array 1382744_at - -
          Affymetrix HT Rat Focus Array 1398573_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700004742.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004743.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004738.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004739.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004745.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004744.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004740.rn.1 TC1700000694.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004741.rn.1 TC1700000694.rn.1 RTA TC
          Rat Expression Set 230 1382744_at - -
          Rat Expression Set 230 1398573_PM_at - -
          Rat Expression Set 230 1398573_at - -
          Rat Expression Set 230 1382744_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795335 - -
          Rat Genome U34 Set rc_AI175753_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21302572 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline