Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528582_m1
          See other TMEM164 GE Assays ›
          Gene Symbol:
          Tmem164
          Gene Name:
          transmembrane protein 164
          Gene Aliases:
          RGD1563286
          Chromosome Location:
          Chr.X: 113947356 - 114110064 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528582_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem164
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109014.1 NP_001102484.1 6 - 7 1025 83
          XM_008773426.1 XP_008771648.1 6 - 7 1054 83
          XM_008773427.1 XP_008771649.1 5 - 6 913 83
          XM_008773428.1 XP_008771650.1 5 - 6 451 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000972.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P680186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117168 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132927 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63374 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180642 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054616 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054616 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054616 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P680186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76059 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28112 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23165 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937385 - -
          Affymetrix Rat Gene 1.0 ST Array 10937388 - -
          Affymetrix Rat Gene 1.0 ST Array 10937390 - -
          Affymetrix Rat Gene 1.0 ST Array 10937386 - -
          Affymetrix Rat Gene 1.0 ST Array 10937389 - -
          Affymetrix Rat Gene 1.0 ST Array 10937387 - -
          Affymetrix Rat Gene 1.1 ST Array 10937382 - -
          Rat Genome 230 2.0 Array 1396478_PM_at - -
          Rat Genome 230 2.0 Array 1374368_at - -
          Rat Genome 230 2.0 Array 1374368_PM_at - -
          Rat Genome 230 2.0 Array 1396478_at - -
          Affymetrix HT Rat Focus Array 1396478_at - -
          Affymetrix HT Rat Focus Array 1374368_at - -
          Affymetrix HT Rat Focus Array 1396478_PM_at - -
          Affymetrix HT Rat Focus Array 1374368_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00006029.rn.1 TC0X00000972.rn.1 RTA TC
          Rat Expression Set 230 1374368_at - -
          Rat Expression Set 230 1374368_PM_at - -
          Rat Expression Set 230 1396478_at - -
          Rat Expression Set 230 1396478_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937382 - -
          Rat Genome U34 Set rc_AI235156_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21016182 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355557 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline