Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528873_m1
          See other VAPB GE Assays ›
          Gene Symbol:
          Vapb
          Gene Name:
          VAMP (vesicle-associated membrane protein)-associated protein B and C
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 171832558 - 171870198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528873_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vapb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021847.3 NP_068619.1 5 - 6 575 128
          XM_006235706.2 XP_006235768.2 5 - 6 749 128
          GenBank mRNA AF086631.1 - 5 - 6 575 128
          BC065576.1 - 5 - 6 760 5598396 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002505.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vapb

          Panther Classification:

          Molecular Function -

          membrane traffic protein membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular calcium ion homeostasis
          ER to Golgi vesicle-mediated transport
          activation of signaling protein activity involved in unfolded protein response
          endoplasmic reticulum organization
          biological_process
          modulation by virus of host morphology or physiology
          endoplasmic reticulum unfolded protein response
          negative regulation by host of viral release from host cell
          positive regulation by host of viral release from host cell
          negative regulation by host of viral genome replication
          positive regulation by host of viral genome replication
          positive regulation of viral genome replication
          negative regulation by virus of viral protein levels in host cell
          COPII-coated vesicle budding
          molecular_function
          microtubule binding
          enzyme binding
          FFAT motif binding
          protein homodimerization activity
          protein heterodimerization activity
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P545129 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152057 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117408 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117408 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492843 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700529157 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13963 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83573 - -
          Agilent Rat cDNA Microarray (G4105A) 700529157 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842628 - -
          Affymetrix Rat Gene 1.0 ST Array 10842626 - -
          Affymetrix Rat Gene 1.0 ST Array 10842629 - -
          Affymetrix Rat Gene 1.0 ST Array 10842630 - -
          Affymetrix Rat Gene 1.0 ST Array 10842627 - -
          Affymetrix Rat Gene 1.0 ST Array 10842632 - -
          Affymetrix Rat Gene 1.0 ST Array 10842631 - -
          Affymetrix Rat Gene 1.0 ST Array 10842625 - -
          Affymetrix Rat Gene 1.1 ST Array 10842624 - -
          Rat Genome 230 2.0 Array 1369597_PM_at - -
          Rat Genome 230 2.0 Array 1390257_at - -
          Rat Genome 230 2.0 Array 1390257_PM_at - -
          Rat Genome 230 2.0 Array 1369597_at - -
          Affymetrix HT Rat Focus Array 1390257_PM_at - -
          Affymetrix HT Rat Focus Array 1369597_at - -
          Affymetrix HT Rat Focus Array 1369597_PM_at - -
          Affymetrix HT Rat Focus Array 1390257_at - -
          Affymetrix Rat Clariom S Assay PSR0300018486.rn.1 TC0300002505.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300018487.rn.1 TC0300002505.rn.1 RTA TC
          Rat Expression Set 230 1369597_at - -
          Rat Expression Set 230 1390257_at - -
          Rat Expression Set 230 1390257_PM_at - -
          Rat Expression Set 230 1369597_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842624 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22148950 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366698 - -
          RatRef-12 Expression BeadChip ILMN_60859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline