Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528937_m1
          See other STX16 GE Assays ›
          Gene Symbol:
          Stx16
          Gene Name:
          syntaxin 16
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 172154742 - 172183699 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528937_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stx16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108610.1 NP_001102080.1 6 - 7 713 86
          XM_006235693.2 XP_006235755.1 8 - 9 1641 86
          XM_006235694.2 XP_006235756.1 7 - 8 1629 86
          XM_006235695.2 XP_006235757.1 8 - 9 1590 86
          XM_006235696.2 XP_006235758.1 7 - 8 1577 86
          GenBank mRNA BC168980.1 - 5 - 6 567 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002509.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          intra-Golgi vesicle-mediated transport
          vesicle fusion
          retrograde transport, endosome to Golgi
          vesicle docking
          Golgi ribbon formation
          SNARE binding
          SNAP receptor activity
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P173345 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025083 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P173345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P173345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P173345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025083 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036815 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842641 - -
          Affymetrix Rat Gene 1.0 ST Array 10842639 - -
          Affymetrix Rat Gene 1.0 ST Array 10842637 - -
          Affymetrix Rat Gene 1.0 ST Array 10842640 - -
          Affymetrix Rat Gene 1.0 ST Array 10842636 - -
          Affymetrix Rat Gene 1.0 ST Array 10842638 - -
          Affymetrix Rat Gene 1.0 ST Array 10842643 - -
          Affymetrix Rat Gene 1.0 ST Array 10842642 - -
          Affymetrix Rat Gene 1.1 ST Array 10842635 - -
          Rat Genome 230 2.0 Array 1367530_at - -
          Rat Genome 230 2.0 Array 1367530_PM_at - -
          Affymetrix HT Rat Focus Array 1367530_PM_at - -
          Affymetrix HT Rat Focus Array 1367530_at - -
          Affymetrix Rat Clariom S Assay PSR0300018511.rn.1 TC0300002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300018510.rn.1 TC0300002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300018509.rn.1 TC0300002509.rn.1 RTA TC
          Rat Expression Set 230 1367530_PM_at - -
          Rat Expression Set 230 1367530_at - -
          Rat Expression Set 230 1367530_PM_at - -
          Rat Expression Set 230 1367530_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842635 - -
          Rat Genome U34 Set rc_AI145853_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21363905 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350357 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline