Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529185_m1
          See other CDK6 GE Assays ›
          Gene Symbol:
          Cdk6
          Gene Name:
          cyclin-dependent kinase 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 27781728 - 27969252 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529185_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdk6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191861.1 NP_001178790.1 5 - 6 696 128
          XM_006236019.2 XP_006236081.1 6 - 7 1160 128
          XM_006236020.2 XP_006236082.1 6 - 7 1094 128
          XM_006236021.1 XP_006236083.1 6 - 7 1036 128
          XM_008762679.1 XP_008760901.1 6 - 7 1011 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002370.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell-matrix adhesion
          hematopoietic progenitor cell differentiation
          type B pancreatic cell development
          protein phosphorylation
          Notch signaling pathway
          response to virus
          regulation of gene expression
          positive regulation of gene expression
          phosphorylation
          hemopoiesis
          T cell differentiation in thymus
          gliogenesis
          cell dedifferentiation
          negative regulation of myeloid cell differentiation
          regulation of erythrocyte differentiation
          negative regulation of osteoblast differentiation
          negative regulation of cell cycle
          positive regulation of fibroblast proliferation
          negative regulation of epithelial cell proliferation
          hematopoietic stem cell differentiation
          negative regulation of cellular senescence
          cyclin-dependent protein serine/threonine kinase activity
          ATP binding
          kinase activity
          cyclin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P113443 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113443 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113443 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P113443 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P405071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45438 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79418 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37112 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10860772 - -
          Affymetrix Rat Gene 1.0 ST Array 10860771 - -
          Affymetrix Rat Gene 1.0 ST Array 10860770 - -
          Affymetrix Rat Gene 1.1 ST Array 10860765 - -
          Rat Genome 230 2.0 Array 1378622_at - -
          Rat Genome 230 2.0 Array 1378622_PM_at - -
          Rat Genome 230 2.0 Array 1375757_PM_at - -
          Rat Genome 230 2.0 Array 1375757_at - -
          Affymetrix Rat Clariom S Assay PSR0400016949.rn.1 TC0400002370.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400016950.rn.1 TC0400002370.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400016952.rn.1 TC0400002370.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400016948.rn.1 TC0400002370.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400016953.rn.1 TC0400002370.rn.1 RTA TC
          Rat Expression Set 230 1375757_PM_at - -
          Rat Expression Set 230 1375757_at - -
          Affymetrix Rat Gene 1.0 ST Array 10860765 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20942573 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374074 - -
          RatRef-12 Expression BeadChip ILMN_58924 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline