Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529322_g1
          See other CD63 GE Assays ›
          Gene Symbol:
          Cd63
          Gene Name:
          Cd63 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 3320250 - 3335581 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529322_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017125.3 NP_058821.1 3 - 4 314 69
          XM_008765024.1 XP_008763246.1 3 - 4 376 69
          XM_008765025.1 XP_008763247.1 3 - 4 579 69
          GenBank mRNA BC063173.1 - 3 - 4 317 6921594 69
          FQ214468.1 - 3 - 4 381 69
          FQ221333.1 - 3 - 4 318 69
          FQ222038.1 - 3 - 4 297 69
          FQ230116.1 - 3 - 4 285 69
          X61654.1 - 3 - 4 314 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000052.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd63

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          cell-matrix adhesion
          negative regulation of epithelial cell migration
          protein transport
          cell migration
          epithelial cell differentiation
          cellular protein localization
          endosome to melanosome transport
          pigmentation
          positive regulation of cell adhesion
          positive regulation of endocytosis
          pigment granule maturation
          pigment cell differentiation
          regulation of vascular endothelial growth factor signaling pathway
          regulation of rubidium ion transport
          positive regulation of integrin-mediated signaling pathway
          protein binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P744495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P744495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P744495 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P744495 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P744495 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700147074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15178 - -
          Agilent Rat cDNA Microarray (G4105A) 700147074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10893261 - -
          Affymetrix Rat Gene 1.0 ST Array 10893265 - -
          Affymetrix Rat Gene 1.0 ST Array 10893262 - -
          Affymetrix Rat Gene 1.0 ST Array 10893264 - -
          Affymetrix Rat Gene 1.0 ST Array 10893259 - -
          Affymetrix Rat Gene 1.0 ST Array 10893260 - -
          Affymetrix Rat Gene 1.0 ST Array 10893266 - -
          Affymetrix Rat Gene 1.0 ST Array 10893263 - -
          Affymetrix Rat Gene 1.1 ST Array 10899951 - -
          Affymetrix Rat Gene 1.1 ST Array 10893258 - -
          Rat Genome 230 2.0 Array 1367709_at - -
          Rat Genome 230 2.0 Array 1367709_PM_at - -
          Affymetrix HT Rat Focus Array 1367709_PM_at - -
          Affymetrix HT Rat Focus Array 1367709_at - -
          Affymetrix Rat Clariom S Assay PSR0700000742.rn.1 TC0700000052.rn.1 RTA TC
          Rat Expression Set 230 1367709_PM_at - -
          Rat Expression Set 230 1367709_at - -
          Affymetrix Rat Gene 1.0 ST Array 10893258 - -
          Rat Genome U34 Set X61654_at - -
          Rat Genome U34 Set rc_AA997844_f_at - -
          Rat Genome U34 Set rc_AA997844_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21640737 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355682 - -
          RatRef-12 Expression BeadChip ILMN_67382 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline