Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529624_g1
          See other PNCK GE Assays ›
          Gene Symbol:
          Pnck
          Gene Name:
          pregnancy up-regulated nonubiquitous CaM kinase
          Gene Aliases:
          Camk1b
          Chromosome Location:
          Chr.X: 157150078 - 157154558 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529624_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pnck
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017275.1 NP_058971.1 9 - 10 1017 66
          XM_006229572.2 XP_006229634.1 9 - 10 849 66
          XM_006229573.2 XP_006229635.1 9 - 10 873 66
          XM_006229574.2 XP_006229636.1 9 - 10 823 66
          XM_006229575.2 XP_006229637.1 9 - 10 827 66
          XM_008773628.1 XP_008771850.1 9 - 10 942 66
          XM_008773629.1 XP_008771851.1 8 - 9 820 66
          GenBank mRNA AB004267.1 - 9 - 10 1017 66
          D86556.1 - 9 - 10 868 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001316.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pnck

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          calmodulin-dependent protein kinase activity
          calmodulin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P118992 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118992 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118992 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069988 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118992 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P604657 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700766030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13174 - -
          Agilent Rat cDNA Microarray (G4105A) 700766030 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940155 - -
          Affymetrix Rat Gene 1.0 ST Array 10940159 - -
          Affymetrix Rat Gene 1.0 ST Array 10940158 - -
          Affymetrix Rat Gene 1.0 ST Array 10940157 - -
          Affymetrix Rat Gene 1.0 ST Array 10940151 - -
          Affymetrix Rat Gene 1.0 ST Array 10940152 - -
          Affymetrix Rat Gene 1.0 ST Array 10940162 - -
          Affymetrix Rat Gene 1.0 ST Array 10940161 - -
          Affymetrix Rat Gene 1.0 ST Array 10940156 - -
          Affymetrix Rat Gene 1.0 ST Array 10940153 - -
          Affymetrix Rat Gene 1.0 ST Array 10940154 - -
          Affymetrix Rat Gene 1.1 ST Array 10940150 - -
          Rat Genome 230 2.0 Array 1387281_PM_a_at - -
          Rat Genome 230 2.0 Array 1387281_a_at - -
          Affymetrix HT Rat Focus Array 1387281_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387281_a_at - -
          Affymetrix Rat Clariom S Assay PSR0X00008144.rn.1 TC0X00001316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00008143.rn.1 TC0X00001316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00008142.rn.1 TC0X00001316.rn.1 RTA TC
          Rat Expression Set 230 1387281_a_at - -
          Rat Expression Set 230 1387281_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940150 - -
          Rat Genome U34 Set D86556_i_at - -
          Rat Genome U34 Set AB004267_g_at - -
          Rat Genome U34 Set AB004267_at - -
          Rat Genome U34 Set AB004267_at - -
          Rat Genome U34 Set D86556_i_at - -
          Rat Genome U34 Set AB004267_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20762432 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357156 - -
          RatRef-12 Expression BeadChip ILMN_66828 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline