Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529640_g1
          See other G6PD GE Assays ›
          Gene Symbol:
          G6pd
          Gene Name:
          glucose-6-phosphate dehydrogenase
          Gene Aliases:
          G6pdx
          Chromosome Location:
          Chr.X: 156274800 - 156293935 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529640_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: G6pd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017006.2 NP_058702.1 11 - 12 1483 72
          GenBank mRNA BC081820.1 - 11 - 12 1396 7108556 72
          X07467.1 - 11 - 12 1409 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001290.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene G6pd

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure
          vasodilation by angiotensin involved in regulation of systemic arterial blood pressure
          glucose metabolic process
          pentose-phosphate shunt
          cholesterol biosynthetic process
          NADP metabolic process
          NADPH regeneration
          NADP biosynthetic process
          glutathione metabolic process
          response to oxidative stress
          pentose-phosphate shunt, oxidative branch
          negative regulation of protein glutathionylation
          response to organic cyclic compound
          pentose biosynthetic process
          substantia nigra development
          response to food
          interleukin-10 production
          interleukin-12 production
          cellular response to oxidative stress
          regulation of multicellular organism growth
          erythrocyte maturation
          regulation of neuron apoptotic process
          response to ethanol
          ribose phosphate biosynthetic process
          erythrocyte development
          glucose 6-phosphate metabolic process
          oxidation-reduction process
          glucose-6-phosphate dehydrogenase activity
          glucose binding
          carbohydrate binding
          identical protein binding
          protein homodimerization activity
          NADP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11770 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11770 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292327 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936054 - -
          Affymetrix Rat Gene 1.0 ST Array 10936051 - -
          Affymetrix Rat Gene 1.0 ST Array 10936049 - -
          Affymetrix Rat Gene 1.0 ST Array 10936052 - -
          Affymetrix Rat Gene 1.0 ST Array 10886407 - -
          Affymetrix Rat Gene 1.0 ST Array 10936053 - -
          Affymetrix Rat Gene 1.0 ST Array 10936050 - -
          Affymetrix Rat Gene 1.1 ST Array 10936048 - -
          Affymetrix Rat Gene 1.1 ST Array 10886406 - -
          Rat Genome 230 2.0 Array 1367856_at - -
          Rat Genome 230 2.0 Array 1367856_PM_at - -
          Affymetrix HT Rat Focus Array 1367856_PM_at - -
          Affymetrix HT Rat Focus Array 1367856_at - -
          Affymetrix Rat Clariom S Assay PSR0X00007627.rn.1 TC0X00001290.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007629.rn.1 TC0X00001290.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007628.rn.1 TC0X00001290.rn.1 RTA TC
          Rat Expression Set 230 1367856_at - -
          Rat Expression Set 230 1367856_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936048 - -
          Affymetrix Rat Gene 1.0 ST Array 10886406 - -
          Rat Genome U34 Set X07467_at - -
          Rat Genome U34 Set X07467_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21469797 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376588 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline