Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529862_m1
          See other PLXNA3 GE Assays ›
          Gene Symbol:
          Plxna3
          Gene Name:
          plexin A3
          Gene Aliases:
          Plxn4, SEX
          Chromosome Location:
          Chr.X: 156363400 - 156379345 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529862_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plxna3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107581.2 NP_001101051.2 32 - 33 5695 85
          XM_006229577.2 XP_006229639.1 32 - 33 5854 85
          XM_006229578.2 XP_006229640.1 32 - 33 6111 85
          XM_008773598.1 XP_008771820.1 31 - 32 7603 85
          XM_008773599.1 XP_008771821.1 31 - 32 7372 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002629.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          axon guidance
          facial nerve structural organization
          trigeminal nerve structural organization
          hippocampus development
          branchiomotor neuron axon guidance
          pyramidal neuron development
          negative regulation of axon extension involved in axon guidance
          negative chemotaxis
          positive regulation of cytoskeleton organization
          semaphorin-plexin signaling pathway
          semaphorin-plexin signaling pathway involved in axon guidance
          semaphorin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21050 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21050 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21050 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21050 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25934 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940350 - -
          Affymetrix Rat Gene 1.0 ST Array 10940356 - -
          Affymetrix Rat Gene 1.0 ST Array 10940342 - -
          Affymetrix Rat Gene 1.0 ST Array 10940332 - -
          Affymetrix Rat Gene 1.0 ST Array 10940333 - -
          Affymetrix Rat Gene 1.0 ST Array 10940338 - -
          Affymetrix Rat Gene 1.0 ST Array 10940357 - -
          Affymetrix Rat Gene 1.0 ST Array 10940337 - -
          Affymetrix Rat Gene 1.0 ST Array 10940354 - -
          Affymetrix Rat Gene 1.0 ST Array 10940355 - -
          Affymetrix Rat Gene 1.0 ST Array 10940329 - -
          Affymetrix Rat Gene 1.0 ST Array 10940336 - -
          Affymetrix Rat Gene 1.0 ST Array 10940340 - -
          Affymetrix Rat Gene 1.0 ST Array 10940343 - -
          Affymetrix Rat Gene 1.0 ST Array 10940346 - -
          Affymetrix Rat Gene 1.0 ST Array 10940331 - -
          Affymetrix Rat Gene 1.0 ST Array 10940325 - -
          Affymetrix Rat Gene 1.0 ST Array 10940353 - -
          Affymetrix Rat Gene 1.0 ST Array 10940339 - -
          Affymetrix Rat Gene 1.0 ST Array 10940345 - -
          Affymetrix Rat Gene 1.0 ST Array 10940351 - -
          Affymetrix Rat Gene 1.0 ST Array 10940328 - -
          Affymetrix Rat Gene 1.0 ST Array 10940352 - -
          Affymetrix Rat Gene 1.0 ST Array 10940348 - -
          Affymetrix Rat Gene 1.0 ST Array 10940326 - -
          Affymetrix Rat Gene 1.0 ST Array 10940349 - -
          Affymetrix Rat Gene 1.0 ST Array 10940330 - -
          Affymetrix Rat Gene 1.0 ST Array 10940347 - -
          Affymetrix Rat Gene 1.0 ST Array 10940327 - -
          Affymetrix Rat Gene 1.0 ST Array 10940341 - -
          Affymetrix Rat Gene 1.0 ST Array 10940335 - -
          Affymetrix Rat Gene 1.0 ST Array 10940334 - -
          Affymetrix Rat Gene 1.0 ST Array 10940344 - -
          Affymetrix Rat Gene 1.1 ST Array 10940324 - -
          Rat Genome 230 2.0 Array 1376139_at - -
          Rat Genome 230 2.0 Array 1377077_PM_at - -
          Rat Genome 230 2.0 Array 1377077_at - -
          Rat Genome 230 2.0 Array 1376139_PM_at - -
          Affymetrix HT Rat Focus Array 1377077_at - -
          Affymetrix HT Rat Focus Array 1377077_PM_at - -
          Affymetrix HT Rat Focus Array 1376139_PM_at - -
          Affymetrix HT Rat Focus Array 1376139_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016240.rn.1 TC0X00002629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016241.rn.1 TC0X00002629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016239.rn.1 TC0X00002629.rn.1 RTA TC
          Rat Expression Set 230 1376139_at - -
          Rat Expression Set 230 1377077_PM_at - -
          Rat Expression Set 230 1376139_PM_at - -
          Rat Expression Set 230 1377077_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940324 - -
          Rat Genome U34 Set rc_AI059099_at - -
          Rat Genome U34 Set rc_AI175628_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21713273 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360444 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline