Search
Search

RGD1307443| Gene Symbol: RGD1307443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197023.1 | NP_001183952.1 | 16 - 17 | 2858 | 82 | |
| XM_006253936.2 | XP_006253998.1 | 16 - 17 | 3841 | 82 | ||
Set Membership: |
|