Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530118_m1
          See other HIST1H2BH GE Assays ›
          Gene Symbol:
          Hist1h2bh
          Gene Name:
          histone cluster 1, H2bh
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 43679925 - 43689311 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530118_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h2bh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107352.1 NP_001100822.1 1 - 2 470 101
          GenBank mRNA BC079458.1 - 1 - 2 388 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001802.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          antibacterial humoral response
          defense response to Gram-positive bacterium
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020269 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P162197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P020269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P162197 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139989 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798441 - -
          Affymetrix Rat Gene 1.0 ST Array 10795190 - -
          Affymetrix Rat Gene 1.0 ST Array 10795191 - -
          Affymetrix Rat Gene 1.0 ST Array 10798496 - -
          Affymetrix Rat Gene 1.0 ST Array 10798489 - -
          Affymetrix Rat Gene 1.0 ST Array 10798498 - -
          Affymetrix Rat Gene 1.1 ST Array 10798440 - -
          Affymetrix Rat Gene 1.1 ST Array 10795189 - -
          Rat Genome 230 2.0 Array 1391301_PM_at - -
          Rat Genome 230 2.0 Array 1390021_PM_at - -
          Rat Genome 230 2.0 Array 1390021_at - -
          Rat Genome 230 2.0 Array 1391301_at - -
          Affymetrix HT Rat Focus Array 1390021_PM_at - -
          Affymetrix HT Rat Focus Array 1390021_at - -
          Affymetrix Rat Clariom S Assay PSR1700012386.rn.1 TC1700001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012385.rn.1 TC1700001802.rn.1 RTA TC
          Rat Expression Set 230 1390021_at - -
          Rat Expression Set 230 1390021_PM_at - -
          Rat Expression Set 230 1391301_at - -
          Rat Expression Set 230 1391301_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795189 - -
          Rat Genome U34 Set rc_AA964055_at - -
          Rat Genome U34 Set rc_AA997499_i_at - -
          Rat Genome U34 Set rc_AI012354_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21720617 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline