Search
Search
| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014009.2 | NP_001014031.2 | 15 - 16 | 1450 | 79 | |
| XM_006253929.2 | XP_006253991.1 | 11 - 12 | 1753 | 79 | ||
| XM_006253930.1 | XP_006253992.1 | 11 - 12 | 1109 | 79 | ||
| XM_008771636.1 | XP_008769858.1 | 11 - 12 | 1593 | 79 | ||
| XM_008771637.1 | XP_008769859.1 | 10 - 11 | 1029 | 79 | ||
| XM_008771638.1 | XP_008769860.1 | 11 - 12 | 1594 | 79 | ||
| XM_008771639.1 | XP_008769861.1 | 11 - 12 | 1208 | 79 | ||
| GenBank mRNA | AY325194.1 | - | 15 - 16 | 1450 | 79 | |
Set Membership: |
|