Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530730_g1
          See other RSL24D1 GE Assays ›
          Gene Symbol:
          Rsl24d1
          Gene Name:
          ribosomal L24 domain containing 1
          Gene Aliases:
          RGD1309784
          Chromosome Location:
          Chr.8: 79792587 - 79801592 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530730_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsl24d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014212.1 NP_001014234.1 4 - 5 376 120
          GenBank mRNA BC062237.1 - 4 - 5 376 6919069 120

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsl24d1

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s)

          ribosome biogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10744 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10744 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97670 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911543 - -
          Affymetrix Rat Gene 1.0 ST Array 10911539 - -
          Affymetrix Rat Gene 1.0 ST Array 10911541 - -
          Affymetrix Rat Gene 1.0 ST Array 10911542 - -
          Affymetrix Rat Gene 1.0 ST Array 10911540 - -
          Affymetrix Rat Gene 1.1 ST Array 10911538 - -
          Rat Genome 230 2.0 Array 1395449_at - -
          Rat Genome 230 2.0 Array 1395449_PM_at - -
          Affymetrix HT Rat Focus Array 1395449_PM_at - -
          Affymetrix HT Rat Focus Array 1395449_at - -
          Affymetrix Rat Clariom S Assay PSR0800007963.rn.1 TC0800001088.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007965.rn.1 TC0800001088.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007962.rn.1 TC0800001088.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007967.rn.1 TC0800001088.rn.1 RTA TC
          Rat Expression Set 230 1395449_PM_at - -
          Rat Expression Set 230 1395449_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911538 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21398520 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368274 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline