Search
Search
| Gene Symbol: Gtf2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053345.1 | NP_445797.1 | 2 - 3 | 178 | 75 | |
| XM_006243380.2 | XP_006243442.1 | 3 - 4 | 328 | 75 | ||
| GenBank mRNA | AF000944.1 | - | 2 - 3 | 178 | 75 | |
| FQ209546.1 | - | 333 | 75 | |||
| FQ212213.1 | - | 3 - 4 | 313 | 75 | ||
| FQ221947.1 | - | 3 - 4 | 319 | 75 | ||
| FQ228685.1 | - | 3 - 4 | 333 | 75 | ||
Set Membership: |
|