Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530876_m1
          See other LILRB3L GE Assays ›
          Gene Symbol:
          Lilrb3l
          Gene Name:
          leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3-like
          Gene Aliases:
          Lilrb3, Pir-A1, Pir-A2, Pirb
          Chromosome Location:
          Chr.1: 63759638 - 63849278 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530876_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lilrb3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037357.2 NP_001032434.1 679 77
          XM_006228032.2 XP_006228094.1 4 - 5 721 77
          XM_006228033.2 XP_006228095.1 4 - 5 721 77
          XM_006228034.2 XP_006228096.1 3 - 4 436 77
          GenBank mRNA AF169636.1 - 670 77
          AF169637.1 - 5 - 6 679 77
          AF169638.1 - 679 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lilrb3l

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P111058 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178353 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112684 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112684 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P111058 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517958 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56680 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16632 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43109 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55133 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84506 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60397 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26463 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100392 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97586 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10703687 - -
          Affymetrix Rat Gene 1.0 ST Array 10703690 - -
          Affymetrix Rat Gene 1.0 ST Array 10703688 - -
          Affymetrix Rat Gene 1.0 ST Array 10703701 - -
          Affymetrix Rat Gene 1.0 ST Array 10703700 - -
          Affymetrix Rat Gene 1.0 ST Array 10703691 - -
          Affymetrix Rat Gene 1.0 ST Array 10703685 - -
          Affymetrix Rat Gene 1.0 ST Array 10703702 - -
          Affymetrix Rat Gene 1.0 ST Array 10703689 - -
          Affymetrix Rat Gene 1.0 ST Array 10703703 - -
          Affymetrix Rat Gene 1.0 ST Array 10703681 - -
          Affymetrix Rat Gene 1.0 ST Array 10703682 - -
          Affymetrix Rat Gene 1.0 ST Array 10703705 - -
          Affymetrix Rat Gene 1.0 ST Array 10703686 - -
          Affymetrix Rat Gene 1.1 ST Array 10703680 - -
          Rat Genome 230 2.0 Array 1370682_PM_at - -
          Rat Genome 230 2.0 Array 1387992_at - -
          Rat Genome 230 2.0 Array 1370682_at - -
          Rat Genome 230 2.0 Array 1370493_a_at - -
          Rat Genome 230 2.0 Array 1370493_PM_a_at - -
          Rat Genome 230 2.0 Array 1387992_PM_at - -
          Affymetrix HT Rat Focus Array 1387992_at - -
          Affymetrix HT Rat Focus Array 1370493_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370682_PM_at - -
          Affymetrix HT Rat Focus Array 1387992_PM_at - -
          Affymetrix HT Rat Focus Array 1370493_a_at - -
          Affymetrix HT Rat Focus Array 1370682_at - -
          Rat Expression Set 230 1370682_PM_at - -
          Rat Expression Set 230 1370493_a_at - -
          Rat Expression Set 230 1370682_at - -
          Rat Expression Set 230 1387992_at - -
          Rat Expression Set 230 1387992_PM_at - -
          Rat Expression Set 230 1370493_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10703680 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22057966 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline