Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01531050_m1
          See other TMEM175 GE Assays ›
          Gene Symbol:
          Tmem175
          Gene Name:
          transmembrane protein 175
          Gene Aliases:
          RGD1309712
          Chromosome Location:
          Chr.14: 2083745 - 2100046 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01531050_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem175
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013991.1 NP_001014013.1 10 - 11 979 74
          XM_006250558.2 XP_006250620.1 10 - 11 1435 74
          XM_008769936.1 XP_008768158.1 8 - 9 1662 74
          XM_008769937.1 XP_008768159.1 7 - 8 900 74
          GenBank mRNA BC079245.1 - 10 - 11 979 7132812 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001233.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem175

          Panther Classification:

          Molecular Function -

          ion channel

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P501365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2564 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P501365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P501365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P501365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501365 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99223 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775027 - -
          Affymetrix Rat Gene 1.0 ST Array 10775021 - -
          Affymetrix Rat Gene 1.0 ST Array 10775025 - -
          Affymetrix Rat Gene 1.0 ST Array 10775019 - -
          Affymetrix Rat Gene 1.0 ST Array 10775026 - -
          Affymetrix Rat Gene 1.0 ST Array 10775017 - -
          Affymetrix Rat Gene 1.0 ST Array 10775022 - -
          Affymetrix Rat Gene 1.0 ST Array 10775024 - -
          Affymetrix Rat Gene 1.0 ST Array 10775023 - -
          Affymetrix Rat Gene 1.0 ST Array 10775020 - -
          Affymetrix Rat Gene 1.0 ST Array 10775018 - -
          Affymetrix Rat Gene 1.1 ST Array 10775016 - -
          Rat Genome 230 2.0 Array 1374109_PM_at - -
          Rat Genome 230 2.0 Array 1392306_PM_at - -
          Rat Genome 230 2.0 Array 1392306_at - -
          Rat Genome 230 2.0 Array 1374109_at - -
          Affymetrix Rat Clariom S Assay PSR1400009067.rn.1 TC1400001233.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009068.rn.1 TC1400001233.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10775016 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374099 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0