Search
Search
| Gene Symbol: Nxf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191731.2 | NP_001178660.2 | 3 - 4 | 485 | 111 | |
| XM_006257271.2 | XP_006257333.1 | 4 - 5 | 570 | 111 | ||
| XM_008773409.1 | XP_008771631.1 | 3 - 4 | 500 | 111 | ||
| XM_008773410.1 | XP_008771632.1 | 4 - 5 | 626 | 111 | ||
| XM_008773411.1 | XP_008771633.1 | 4 - 5 | 623 | 111 | ||
| XM_008773412.1 | XP_008771634.1 | 4 - 5 | 503 | 111 | ||
Set Membership: |
|