Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01531762_m1
          See other SLC12A3 GE Assays ›
          Gene Symbol:
          Slc12a3
          Gene Name:
          solute carrier family 12 (sodium/chloride transporter), member 3
          Gene Aliases:
          NCC, TSC
          Chromosome Location:
          Chr.19: 11106033 - 11144674 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01531762_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc12a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019345.3 NP_062218.3 24 - 25 2884 55
          XM_008772323.1 XP_008770545.1 25 - 26 3001 55
          XM_008772324.1 XP_008770546.1 25 - 26 2998 55
          XM_008772325.1 XP_008770547.1 25 - 26 2998 55
          XM_008772326.1 XP_008770548.1 24 - 25 2914 55
          XM_008772327.1 XP_008770549.1 25 - 26 2995 55
          XM_008772328.1 XP_008770550.1 25 - 26 2992 55
          GenBank mRNA U10097.2 - 24 - 25 2884 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001008.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc12a3

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          sodium ion transport
          chloride transport
          sodium ion transmembrane transport
          renal sodium ion absorption
          chloride transmembrane transport
          transporter activity
          sodium ion transmembrane transporter activity
          cation:chloride symporter activity
          sodium:chloride symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015612 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72384 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015612 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015612 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205637 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701716541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66010 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21985 - -
          Agilent Rat cDNA Microarray (G4105A) 700296049 - -
          Agilent Rat cDNA Microarray (G4105A) 701716541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809345 - -
          Affymetrix Rat Gene 1.0 ST Array 10809348 - -
          Affymetrix Rat Gene 1.0 ST Array 10809343 - -
          Affymetrix Rat Gene 1.0 ST Array 10809354 - -
          Affymetrix Rat Gene 1.0 ST Array 10809353 - -
          Affymetrix Rat Gene 1.0 ST Array 10809342 - -
          Affymetrix Rat Gene 1.0 ST Array 10809349 - -
          Affymetrix Rat Gene 1.0 ST Array 10809341 - -
          Affymetrix Rat Gene 1.0 ST Array 10809359 - -
          Affymetrix Rat Gene 1.0 ST Array 10809347 - -
          Affymetrix Rat Gene 1.0 ST Array 10809339 - -
          Affymetrix Rat Gene 1.0 ST Array 10809364 - -
          Affymetrix Rat Gene 1.0 ST Array 10809338 - -
          Affymetrix Rat Gene 1.0 ST Array 10809360 - -
          Affymetrix Rat Gene 1.0 ST Array 10809365 - -
          Affymetrix Rat Gene 1.0 ST Array 10809357 - -
          Affymetrix Rat Gene 1.0 ST Array 10809351 - -
          Affymetrix Rat Gene 1.0 ST Array 10809340 - -
          Affymetrix Rat Gene 1.0 ST Array 10809361 - -
          Affymetrix Rat Gene 1.0 ST Array 10809363 - -
          Affymetrix Rat Gene 1.0 ST Array 10809352 - -
          Affymetrix Rat Gene 1.0 ST Array 10809350 - -
          Affymetrix Rat Gene 1.0 ST Array 10809358 - -
          Affymetrix Rat Gene 1.0 ST Array 10809362 - -
          Affymetrix Rat Gene 1.0 ST Array 10809344 - -
          Affymetrix Rat Gene 1.0 ST Array 10809355 - -
          Affymetrix Rat Gene 1.0 ST Array 10809356 - -
          Affymetrix Rat Gene 1.1 ST Array 10809337 - -
          Rat Genome 230 2.0 Array 1387230_at - -
          Rat Genome 230 2.0 Array 1387230_PM_at - -
          Affymetrix HT Rat Focus Array 1387230_at - -
          Affymetrix HT Rat Focus Array 1387230_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900008121.rn.1 TC1900001008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900008125.rn.1 TC1900001008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900008119.rn.1 TC1900001008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900008123.rn.1 TC1900001008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900008126.rn.1 TC1900001008.rn.1 RTA TC
          Rat Expression Set 230 1387230_at - -
          Rat Expression Set 230 1387230_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809337 - -
          Rat Genome U34 Set U10097_at - -
          Rat Genome U34 Set rc_AA965132_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21294288 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354820 - -
          RatRef-12 Expression BeadChip ILMN_68583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline