Search
Search

Ppp1r18| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126287.2 | NP_001119759.2 | 2 - 3 | 1618 | 87 | |
| XM_006255952.2 | XP_006256014.1 | 1 - 2 | 3342 | 87 | ||
| XM_006255953.2 | XP_006256015.1 | 2 - 3 | 2410 | 87 | ||
| GenBank mRNA | BC161960.1 | - | 2 - 3 | 1618 | 9027646 | 87 |
| FQ227692.1 | - | 1 - 2 | 159 | 87 | ||
| FQ232122.1 | - | 1 - 2 | 135 | 87 | ||
Set Membership: |
|