Search
Search

Zfp385a| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109470.2 | NP_001102940.1 | 2 - 3 | 181 | 59 | |
| NM_001135088.1 | NP_001128560.1 | 2 - 3 | 333 | 59 | ||
| NM_001135089.1 | NP_001128561.1 | 2 - 3 | 232 | 59 | ||
| XM_006242445.2 | XP_006242507.1 | 1 - 2 | 358 | 59 | ||
| XM_006242446.2 | XP_006242508.1 | 2 - 3 | 538 | 59 | ||
| GenBank mRNA | BC166729.1 | - | 2 - 3 | 333 | 7934026 | 59 |
Set Membership: |
|