Search
Search
| Gene Symbol: Ccdc65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014203.1 | NP_001014225.1 | 2 - 3 | 428 | 71 | |
| XM_006257362.2 | XP_006257424.1 | 2 - 3 | 494 | 71 | ||
| GenBank mRNA | BC083683.1 | - | 2 - 3 | 428 | 7135919 | 71 |
Set Membership: |
|