Search
Search
| Gene Symbol: Copz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108117.1 | NP_001101587.1 | 5 - 6 | 366 | 111 | |
| GenBank mRNA | FQ226451.1 | - | 5 - 6 | 111 | ||
| FQ234671.1 | - | 5 - 6 | 342 | 111 | ||
Set Membership: |
|