Search
Search

Nckap1l| Gene Symbol: Nckap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108119.1 | NP_001101589.1 | 18 - 19 | 1908 | 68 | |
| XM_008765753.1 | XP_008763975.1 | 18 - 19 | 1964 | 68 | ||
Set Membership: |
|