Search
Search

LOC680590| Gene Symbol: LOC680590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036618.2 | 7 - 8 | 304 | 78 | ||
| GenBank mRNA | CK597145.1 | - | 8 - 9 | 536 | 78 | |
| CK652853.1 | - | 7 - 8 | 296 | 78 | ||
Set Membership: |
|