Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01532650_m1
          See other FAIM2 GE Assays ›
          Gene Symbol:
          Faim2
          Gene Name:
          Fas apoptotic inhibitory molecule 2
          Gene Aliases:
          Lfg, NMP35
          Chromosome Location:
          Chr.7: 141159749 - 141185567 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01532650_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Faim2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144756.2 NP_653357.1 11 - 12 881 90
          GenBank mRNA AF044201.1 - 11 - 12 830 90
          BC087606.1 - 11 - 12 881 7314068 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003885.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Faim2

          Gene Ontology Categories:

          Function(s)

          response to ischemia
          apoptotic process
          central nervous system development
          cerebellum development
          cerebellar Purkinje cell layer development
          cerebellar granular layer development
          cerebellar Purkinje cell differentiation
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          negative regulation of apoptotic signaling pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P422523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48949 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118741 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P422523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P422523 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P422523 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P422523 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3910 - -
          Agilent Rat cDNA Microarray (G4105A) 700766571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907169 - -
          Affymetrix Rat Gene 1.0 ST Array 10907167 - -
          Affymetrix Rat Gene 1.0 ST Array 10907168 - -
          Affymetrix Rat Gene 1.0 ST Array 10907174 - -
          Affymetrix Rat Gene 1.0 ST Array 10907171 - -
          Affymetrix Rat Gene 1.0 ST Array 10907170 - -
          Affymetrix Rat Gene 1.0 ST Array 10907175 - -
          Affymetrix Rat Gene 1.0 ST Array 10907172 - -
          Affymetrix Rat Gene 1.0 ST Array 10907166 - -
          Affymetrix Rat Gene 1.0 ST Array 10907177 - -
          Affymetrix Rat Gene 1.0 ST Array 10907173 - -
          Affymetrix Rat Gene 1.0 ST Array 10907176 - -
          Affymetrix Rat Gene 1.1 ST Array 10907165 - -
          Rat Genome 230 2.0 Array 1370625_PM_at - -
          Rat Genome 230 2.0 Array 1370625_at - -
          Affymetrix HT Rat Focus Array 1370625_PM_at - -
          Affymetrix HT Rat Focus Array 1370625_at - -
          Affymetrix Rat Clariom S Assay PSR0700031332.rn.1 TC0700003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031331.rn.1 TC0700003885.rn.1 RTA TC
          Rat Expression Set 230 1370625_at - -
          Rat Expression Set 230 1370625_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907165 - -
          Rat Genome U34 Set AF044201_at - -
          Rat Genome U34 Set AF044201_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22338751 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353723 - -
          RatRef-12 Expression BeadChip ILMN_57062 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline