Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01532689_m1
          See other CBX5 GE Assays ›
          Gene Symbol:
          Cbx5
          Gene Name:
          chromobox homolog 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 144819727 - 144865014 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01532689_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cbx5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106797.3 NP_001100267.1 5 - 6 616 100
          XM_006242391.2 XP_006242453.1 5 - 6 680 100
          XM_006242393.2 XP_006242455.1 4 - 5 575 100
          XM_008765707.1 XP_008763929.1 4 - 5 589 100
          GenBank mRNA BC166908.1 - 5 - 6 617 6889335 100
          BC168739.1 - 5 - 6 617 6918617 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003992.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of transcription, DNA-templated
          molecular_function
          chromatin binding
          protein binding, bridging
          methylated histone binding
          protein homodimerization activity
          histone deacetylase binding
          repressing transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1019537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019537 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019537 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1019537 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1019537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907666 - -
          Affymetrix Rat Gene 1.0 ST Array 10907667 - -
          Affymetrix Rat Gene 1.0 ST Array 10907668 - -
          Affymetrix Rat Gene 1.0 ST Array 10907669 - -
          Affymetrix Rat Gene 1.1 ST Array 10907665 - -
          Rat Genome 230 2.0 Array 1373885_at - -
          Rat Genome 230 2.0 Array 1373885_PM_at - -
          Affymetrix HT Rat Focus Array 1373885_at - -
          Affymetrix HT Rat Focus Array 1373885_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700032293.rn.1 TC0700003992.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032294.rn.1 TC0700003992.rn.1 RTA TC
          Rat Expression Set 230 1373885_PM_at - -
          Rat Expression Set 230 1373885_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907665 - -
          Rat Genome U34 Set rc_AA893788_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline