Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01532810_m1
          See other FKBP11 GE Assays ›
          Gene Symbol:
          Fkbp11
          Gene Name:
          FK506 binding protein 11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 140398253 - 140402657 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01532810_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013105.2 NP_001013123.2 5 - 6 483 68
          XM_006257344.2 XP_006257406.1 4 - 5 1388 68
          XM_008765695.1 XP_008763917.1 4 - 5 386 68
          XM_008765696.1 XP_008763918.1 4 - 5 398 68
          XM_008765697.1 XP_008763919.1 4 - 5 375 68
          GenBank mRNA BC084709.1 - 3 - 4 1072 7301886 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700004069.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbp11

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          FK506 binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P192543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185234 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192543 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P192543 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192543 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906937 - -
          Affymetrix Rat Gene 1.0 ST Array 10906936 - -
          Affymetrix Rat Gene 1.0 ST Array 10906935 - -
          Affymetrix Rat Gene 1.0 ST Array 10906940 - -
          Affymetrix Rat Gene 1.0 ST Array 10906938 - -
          Affymetrix Rat Gene 1.0 ST Array 10906939 - -
          Affymetrix Rat Gene 1.0 ST Array 10906941 - -
          Affymetrix Rat Gene 1.0 ST Array 10906943 - -
          Affymetrix Rat Gene 1.1 ST Array 10906934 - -
          Rat Genome 230 2.0 Array 1372653_PM_at - -
          Rat Genome 230 2.0 Array 1372653_at - -
          Affymetrix HT Rat Focus Array 1372653_at - -
          Affymetrix HT Rat Focus Array 1372653_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700033581.rn.1 TC0700004069.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033584.rn.1 TC0700004069.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700033582.rn.1 TC0700004069.rn.1 RTA TC
          Rat Expression Set 230 1372653_at - -
          Rat Expression Set 230 1372653_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10906934 - -
          Rat Genome U34 Set rc_AA875098_at - -
          Rat Genome U34 Set rc_AI045572_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21959604 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline