Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533109_m1
          See other SLC11A2 GE Assays ›
          Gene Symbol:
          Slc11a2
          Gene Name:
          solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2
          Gene Aliases:
          Dmt1, Nramp2
          Chromosome Location:
          Chr.7: 142025812 - 142062996 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533109_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc11a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013173.2 NP_037305.2 4 - 5 403 67
          XM_006242309.2 XP_006242371.1 4 - 5 528 67
          XM_006242310.2 XP_006242372.1 4 - 5 529 67
          XM_006242311.2 XP_006242373.1 4 - 5 438 67
          XM_006242312.2 XP_006242374.1 4 - 5 433 67
          GenBank mRNA AF008439.1 - 4 - 5 414 67
          AF029757.1 - 3 - 4 310 67
          BC097338.1 - 3 - 4 330 7460406 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003905.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc11a2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transition metal ion transport
          response to hypoxia
          detection of oxygen
          porphyrin-containing compound metabolic process
          porphyrin-containing compound biosynthetic process
          heme biosynthetic process
          cobalt ion transport
          copper ion transport
          iron ion transport
          manganese ion transport
          cellular copper ion homeostasis
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          learning or memory
          response to iron ion
          response to manganese ion
          response to lead ion
          nickel cation transport
          vanadium ion transport
          ferrous iron transport
          lead ion transport
          proton transport
          cellular response to oxidative stress
          iron ion transmembrane transport
          copper ion transmembrane transport
          nickel cation transmembrane transport
          response to cadmium ion
          dendrite morphogenesis
          erythrocyte development
          multicellular organismal iron ion homeostasis
          cadmium ion transmembrane transport
          ferrous iron import
          cellular response to iron ion
          cellular response to tumor necrosis factor
          manganese ion transmembrane transport
          cellular response to hypoxia
          zinc II ion transmembrane transport
          cation transmembrane transport
          hydrogen ion transmembrane transport
          copper ion import into cell
          copper ion transmembrane transporter activity
          iron ion transmembrane transporter activity
          manganese ion transmembrane transporter activity
          zinc ion transmembrane transporter activity
          iron ion binding
          copper ion binding
          zinc ion binding
          hydrogen ion transmembrane transporter activity
          calcium ion transmembrane transporter activity
          cadmium ion transmembrane transporter activity
          cobalt ion transmembrane transporter activity
          ferrous iron transmembrane transporter activity
          lead ion transmembrane transporter activity
          nickel cation transmembrane transporter activity
          vanadium ion transmembrane transporter activity
          solute:proton symporter activity
          nickel cation binding
          inorganic cation transmembrane transporter activity
          manganese ion binding
          cadmium ion binding
          cobalt ion binding
          chromium ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P482658 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P102430 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P102433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98024 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P482658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P482658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P482658 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102433 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482658 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P294711 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701032480 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700931521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13241 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22927 - -
          Agilent Rat cDNA Microarray (G4105A) 700931521 - -
          Agilent Rat cDNA Microarray (G4105A) 700915582 - -
          Agilent Rat cDNA Microarray (G4105A) 701032480 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907253 - -
          Affymetrix Rat Gene 1.0 ST Array 10907261 - -
          Affymetrix Rat Gene 1.0 ST Array 10907260 - -
          Affymetrix Rat Gene 1.0 ST Array 10907256 - -
          Affymetrix Rat Gene 1.0 ST Array 10907246 - -
          Affymetrix Rat Gene 1.0 ST Array 10907262 - -
          Affymetrix Rat Gene 1.0 ST Array 10907258 - -
          Affymetrix Rat Gene 1.0 ST Array 10907254 - -
          Affymetrix Rat Gene 1.0 ST Array 10907259 - -
          Affymetrix Rat Gene 1.0 ST Array 10907247 - -
          Affymetrix Rat Gene 1.0 ST Array 10907248 - -
          Affymetrix Rat Gene 1.0 ST Array 10907255 - -
          Affymetrix Rat Gene 1.0 ST Array 10907257 - -
          Affymetrix Rat Gene 1.0 ST Array 10907251 - -
          Affymetrix Rat Gene 1.0 ST Array 10907249 - -
          Affymetrix Rat Gene 1.0 ST Array 10907250 - -
          Affymetrix Rat Gene 1.0 ST Array 10907252 - -
          Affymetrix Rat Gene 1.1 ST Array 10907243 - -
          Rat Genome 230 2.0 Array 1367877_at - -
          Rat Genome 230 2.0 Array 1388059_PM_a_at - -
          Rat Genome 230 2.0 Array 1388059_a_at - -
          Rat Genome 230 2.0 Array 1367877_PM_at - -
          Affymetrix HT Rat Focus Array 1388059_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388059_a_at - -
          Affymetrix HT Rat Focus Array 1367877_PM_at - -
          Affymetrix HT Rat Focus Array 1367877_at - -
          Affymetrix Rat Clariom S Assay PSR0700031505.rn.1 TC0700003905.rn.1 RTA TC
          Rat Expression Set 230 1388059_a_at - -
          Rat Expression Set 230 1367877_at - -
          Rat Expression Set 230 1388059_PM_a_at - -
          Rat Expression Set 230 1367877_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907243 - -
          Rat Genome U34 Set rc_AA892390_s_at - -
          Rat Genome U34 Set AF008439_g_at - -
          Rat Genome U34 Set AF008439_at - -
          Rat Genome U34 Set rc_AA819678_s_at - -
          Rat Genome U34 Set rc_AI177045_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21060590 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368699 - -
          RatRef-12 Expression BeadChip ILMN_49572 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline